Snap Caps mit Rubber/PTFE Septum, rot, zu 12x32mm Vial OMNILAB 0 1 * 100 ST
Catalog # BROW153326
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Snap Caps mit Rubber/PTFE Septum, rot, zu 12x32mm Vial OMNILAB 0 1 * 100 ST
Catalog # BROW153326
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Labelling tapes, temperature resistant
Supplier: PRECISION DYNAMICS TIMEM
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Avantor SterilEnz®-II/AT: pre-gasketed sanitary fittings for single-use systems
Supplier: Avantor Fluid Handling