Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cork stoppers

Cork stoppers

27 mm, 36 mm, 40 mm
Catalog # HECH41594040
Cork stoppers
Catalog # HECH41594040
Cork stoppers
Cork stoppers

27 mm, 36 mm, 40 mm

Price
13.020,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Cork stoppers designed for medical applications.

Specifications
  • Ø bottom:
    36 mm
  • Ø top:
    40 mm
  • Height:
    27 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.