Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Buffers, concentrated solutions, Fluka™

10,00, ±0.05, sodium tetraborate/sodium hydroxide solution, 500 ml, Ampoule, 6
Catalog # 38749-6X1EA
Supplier:   Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Catalog # 38749-6X1EA
CAS Number:  
Buffers, concentrated solutions, Fluka™

10,00, ±0.05, sodium tetraborate/sodium hydroxide solution, 500 ml, Ampoule, 6

CAS Number:  
Price
1.830,00 kr
Log in for availability
1 * 6 Items
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Ampoule
  • Pk:
    6
  • pH accuracy:
    ±0.05
  • Final volume:
    500 ml
  • pH value:
    10,00
  • Contents:
    sodium tetraborate/sodium hydroxide solution

Specifications