Buffers, concentrated solutions, Fluka™
4,00, ±0.02, citric acid/sodium hydroxide/sodium chloride solution, 500 ml, Ampoule, 6
Supplier: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Catalog # 38743-6X1EA
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Ampoule
- Pk:6
- pH accuracy:±0.02
- Final volume:500 ml
- pH value:4,00
- Contents:citric acid/sodium hydroxide/sodium chloride solution



