Buffers, concentrated solutions, Fluka™
1,00, ±0.02, hydrochloric acid/potassium chloride, 500 ml, Ampoule, 1
Supplier: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Catalog # 38740-1EA
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Ampoule
- Pk:1
- pH accuracy:±0.02
- Final volume:500 ml
- pH value:1,00
- Contents:hydrochloric acid/potassium chloride
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



