Order Entry
Denmark
Orders LinkContactUsLinkComponent
Buffers, concentrated solutions, Fluka™

Buffers, concentrated solutions, Fluka™

13,00, ±0.05, glycine/sodium hydroxide/sodium chloride solution, 500 ml, Ampoule, 1
Catalog # 38752-1EA
Supplier:   Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Catalog # 38752-1EA
CAS Number:  
Buffers, concentrated solutions, Fluka™

13,00, ±0.05, glycine/sodium hydroxide/sodium chloride solution, 500 ml, Ampoule, 1

CAS Number:  
Price
278,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • pH accuracy:
    ±0.05
  • Final volume:
    500 ml
  • pH value:
    13,00
  • Contents:
    glycine/sodium hydroxide/sodium chloride solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.