Buffers, concentrated solutions, Fluka™
12,00, ±0.05, di-sodium hydrogen phosphate/sodium hydroxide solution, 500 ml, Ampoule, 1
Supplier: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Catalog # 38751-1EA
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Ampoule
- Pk:1
- pH accuracy:±0.05
- Final volume:500 ml
- pH value:12,00
- Contents:di-sodium hydrogen phosphate/sodium hydroxide solution



