Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium chloride ≥98%, TECHNICAL

Sodium chloride ≥98%, TECHNICAL

Plastic bucket, 25 kg
Catalog # 27788.460
Supplier:   VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Catalog # 27788.460
Supplier:  VWR Chemicals
CAS Number:  7647-14-5
Sodium chloride ≥98%, TECHNICAL
Sodium chloride ≥98%, TECHNICAL

Plastic bucket, 25 kg

CAS Number:  7647-14-5
Price
1.770,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Saline, NaCl, Sea salt, Halite
  • CAS Number:7647-14-5
  • Formula:NaCl
  • MDL Number:MFCD00003477
  • Molecular Weight:58.44 g/mol
  • Merck Index:14,08599
  • Boiling Pt:1413 °C (1013 hPa)
  • Melting Pt:801 °C
  • Density:2.16 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-598-3
Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Appearance:
    Colourless crystals

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.