Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stearic acid

Stearic acid

Glass bottle for solids, 500 mg
Catalog # CAYM10011298-500
Supplier:   Cayman Chemical
Stearic acid
Catalog # CAYM10011298-500
Supplier:  Cayman Chemical
CAS Number:  57-11-4
Stearic acid
Stearic acid

Glass bottle for solids, 500 mg

CAS Number:  57-11-4
Price
309,00 kr
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Boiling Pt:361 °C (1013 hPa)
  • Melting Pt:67 °C
  • Density:0.94 g/cm³ (20 °C)
  • Flash Pt:196 °C (closed cup)
  • Storage Temperature:Refrigerator
  • EINECS:200-313-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.