Apigenin
Glass bottle for solids, 500 mg
Catalog # CAYM10010275-500
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
- CAS Number:520-36-5
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00006831
- Molecular Weight:270.24 g/mol
- EINECS:208-292-3
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



