Order Entry
Denmark
Orders LinkContactUsLinkComponent
Chromatography columns, Flex-Column®, KIMBLE®

Chromatography columns, Flex-Column®, KIMBLE®

10 mm, 200 mm, 16 ml
Catalog # SCERSP420401-1020
Supplier:   DWK Life Sciences
Chromatography columns, Flex-Column®, KIMBLE®
Catalog # SCERSP420401-1020
Supplier:  DWK Life Sciences
Chromatography columns, Flex-Column®, KIMBLE®
Chromatography columns, Flex-Column®, KIMBLE®

10 mm, 200 mm, 16 ml

Price
676,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These economical columns are constructed of polypropylene reservoirs and column outlets permanently connected to a borosilicate glass barrel, molded PP end caps, and HDPE bed supports. They also feature Luer lock inlet and outlet fittings for better connections.

  • Easy connection to any low pressure chromatography system
  • Ideal for use with gel filtration, ion exchange, affinity and adsorption media

PP packing reservoir funnels are for use in initial column packing or as small buffer reservoirs. Glass packing reservoirs are ideal for packing gel slurries into clumns or acting as a buffer reservoir in gravity flow chromatography. Glass reservoirs include a PP upper cap with male Luer lock fitting.

Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. Flow adapters are constructed with a PTFE eluant cannula, a Delrin® body, a Viton® O-ring seal and a 20 µm PE bed support.

Ordering information: Each flow adapter includes one replacment bed support; additional replacements are also available.

Caution: Pressurised glassware should always be used with adequate safety shielding.

Specifications
  • Ø int.:
    10 mm
  • Capacity:
    16 ml
  • Length:
    200 mm
  • Seq:
    14
  • Cat. No.:
    SCERSP420401-1020
  • Max. pressure:
    1,0 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.