Order Entry
Denmark
Orders LinkContactUsLinkComponent
Transport boxes, Securipack

Transport boxes, Securipack

1 L, Transport box for samples, load capacity 5,0 kg, 220×220×360 mm, 140×140×230 mm
Catalog # CORTFABRSECU1G1
Supplier:   E3 Cortex
Transport boxes, Securipack
Catalog # CORTFABRSECU1G1
Supplier:  E3 Cortex
Transport boxes, Securipack
Transport boxes, Securipack

1 L, Transport box for samples, load capacity 5,0 kg, 220×220×360 mm, 140×140×230 mm

Price
769,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

This Securipack packages are ready for use in the transport of samples of hazardous materials, except for Classes 7 and 6.2.

  • Very simple to use, this package eliminates risk posed by error or oversight
  • Mechanical high strength, in particular in the stacking by the juxtaposition of double walls on all the faces
  • High absorption power of the shocks on all the faces, guaranteeing a perfect protection in case of an accidental fall

Universal application for products regulated except for the radioactive and infectious products.

Protects against the following risks: It is a good oxidiser; non flammable and non toxic; corrosive

Ordering information: Supplied with 1 case (sheath), 2 crossed envelopes dressed in a foam thickness, 1 pocket of metallic or plastic retention and 1 sluggish absorbing product. Interior packaging and hazard class label may be supplied upon request.

Specifications
  • W×D×H ext.:
    220×220×360 mm
  • Capacity:
    1 L
  • Description:
    Transport box for samples, load capacity 5,0 kg
  • W×D×H int.:
    140×140×230 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.