Order Entry
Denmark
Orders LinkContactUsLinkComponent
PIG® leak diverter quick-deploy kit

PIG® leak diverter quick-deploy kit

Leak diverter quick-deploy kit, 760×760 mm
Catalog # NEWPTLS461-YW
Supplier:   NEW PIG
PIG® leak diverter quick-deploy kit
Catalog # NEWPTLS461-YW
Supplier:  NEW PIG
PIG® leak diverter quick-deploy kit

Leak diverter quick-deploy kit, 760×760 mm

Price
2.610,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Quick deploying leak diverter springs open fast to catch roof leaks. Simply pop it open.

  • Superfast to control leaks and protect people, inventory, and equipment
  • Hangs easily in problem spots to catch nuisance leaks and channel them to a floor drain or collection container
  • Nonpermeable tarp is vinyl coated for strength and durability; resists punctures, UV and mildew for long life
  • Pinched corners help funnel liquid toward center for better drainage
  • Connect flexible PVC drain hose (included) to channel liquid to floor drain or collection container

3/4" drain fitting accepts longer hoses if necessary. Flow rate of 12,6 litres per minute when used with a 1/2" ID hose.

 : Kit includes leak diverter, quick connect drain fitting (Ø 1905 mm) and hose of length 3 m.

Specifications
  • Description:
    Leak diverter quick-deploy kit
  • L×W:
    760×760 mm
  • Seq:
    02