Nitrate Standard for IC 1 * 100 mL
Catalog # SUPL74246-100ML
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Qualitative filter paper, standard grades MN 615 and 615 1/4, circles, sheets and folded filters
Supplier: MACHEREY-NAGEL
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Glycine analytical reagent, Millipore®
Supplier: Merck
Sterile-venting membrane screw caps, DURAN®
Supplier: DWK Life Sciences
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sulphuric acid 95-97%, EMSURE® ISO for analysis, Supelco®
Supplier: Merck
Test strips, Peroxidase, MQuant®, Supelco®
Supplier: Merck
Wipes for general applications, KIMTECH PURE*
Supplier: KIMBERLY CLARK
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Mixed cellulose ester membranes, non-sterile, 0,45 µm, Whatman™
Supplier: Whatman products (Cytiva)
Lithium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
Bromate standard for IC, TraceCERT®, Supelco®
Supplier: Merck