Order Entry
Denmark
ContactUsLinkComponent
Humidty Ethernet probe 2% with display, incl. wall brack and calibration protocol Protection class: IP54 Measuring rate: 2sec'24h Please notice: Not for condensing atmosphere. For continuous application in high humidty (>80 %RH at #30 C for >12 h, 6 1 * 1 items
Catalog # 620-1627
Supplier:  Testo
Humidty Ethernet probe 2% with display, incl. wall brack and calibration protocol Protection class: IP54 Measuring rate: 2sec'24h Please notice: Not for condensing atmosphere. For continuous application in high humidty (>80 %RH at #30 C for >12 h, 6 1 * 1 items
Catalog # 620-1627
Supplier:  Testo
Frequently Bought Together