Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thermal desorber tubes
Thermal desorber tubes
Thermal desorber tube, with Chromasorb™ 60/80 sorbent
Katalog # PERCN9307055
Leverandør:   PerkinElmer
Thermal desorber tubes
Katalog # PERCN9307055
Leverandør:  PerkinElmer
Thermal desorber tubes
Thermal desorber tube, with Chromasorb™ 60/80 sorbent
Price
10.770,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These desorber tubes are offered with a variety of sorbent packing materials for many GC applications including indoor and outdoor air monitoring, analysis of flavors and fragrances and the analysis of outgassing from packaging, polymers, pharmaceuticals and semi conductor material.
  • Each tube maintains its unique serial number which is etched for easy identification
  • With plastic end caps for short-term storage
Specifikationer
  • Description:
    Thermal desorber tube, with Chromasorb™ 60/80 sorbent
  • Seq:
    05
Anbefalinger