Order Entry
Denmark
Orders LinkContactUsLinkComponent
Absorption granules - Universal coarse grain

Absorption granules - Universal coarse grain

Universal Coarse Grain absorption granules, 1 - 3 mm, 16 litres/sack, 3×20 kg/sack
Catalog # MSGA18468
Supplier:   asecos
Absorption granules - Universal coarse grain
Catalog # MSGA18468
Supplier:  asecos
Absorption granules - Universal coarse grain
Absorption granules - Universal coarse grain

Universal Coarse Grain absorption granules, 1 - 3 mm, 16 litres/sack, 3×20 kg/sack

Price
715,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Absorption granules for oil, alkalis, grease, motor fuels, varnishes.

  • For outdoor and indoor use
  • Officially tested and approved for use on traffic areas
  • 20 kg sack binds up to 16 litres of heating oil
  • Suitable for solid surfaces
  • Even with total saturation of the granules, anti-slipping is guaranteed
Specifications
  • Description:
    Universal Coarse Grain absorption granules, 1 - 3 mm
  • Absorbency:
    16 litres/sack
  • Packed:
    3×20 kg/sack