Order Entry
Denmark
Orders LinkContactUsLinkComponent
Universal mats
Universal mats
Universalmatta, kraftig, 100 mattor/paket, 400×460 mm
Catalog # MSGA9951
Supplier:   asecos
Universal mats
Catalog # MSGA9951
Supplier:  asecos
Universal mats
Universal mats
Universalmatta, kraftig, 100 mattor/paket, 400×460 mm
Price
1.160,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Universal mats for safe absorption of oil, water, cooling liquids and non-aggressive chemicals.
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Mats are pre-cut: only take sufficient material as required
Specifications
  • Description:
    Universalmatta, kraftig
  • L×W:
    400×460 mm
  • Packed:
    100 mattor/paket