Magnetic surface temperature probes
Magnetic surface probe, max. 250 °C (for TFN 520), Lemosa connection, 1 m (Silicone cable), TPN 912, −50...+250 °C
Catalog # EBRO1341-0644
Supplier: EBRO
Jump to:
Product Details & Documents
These probes are used for surface temperature measurement on an extended period of ferrous containing compounds, e.g. heating plates, heating tubes, pumps, flushing tools, motors and turbines.
Specifications
- Range:−50...+250 °C
- Description:Magnetic surface probe, max. 250 °C (for TFN 520), Lemosa connection, 1 m (Silicone cable)
- Seq:2
- Type:TPN 912
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



