Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnels with GL thread or ground joint

Funnels with GL thread or ground joint

20 mm
Catalog # BOHLD738-22
Supplier:   Bohlender
Funnels with GL thread or ground joint
Catalog # BOHLD738-22
Supplier:  Bohlender
Funnels with GL thread or ground joint
Funnels with GL thread or ground joint

20 mm

Price
1.820,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Powder funnels, made of borosilicate glass with outlet pipe of PTFE, connection with GL screw caps of PPS or ground joint.

  • The glass funnel can be fixed in any position
  • Used for reaction vessels with GL threaded connections or grounding sleeves
  • Temperature resistance −20 to +200 °C
  • Universal chemical resistance
Specifications
  • Stem O-Ø:
    20 mm
  • Cat. No.:
    BOHLD738-22