Automatic capillary viscometers, ViscoSystem® AVC 370
Product Details & Documents
ViscoSystem AVC 370 combines a high degree of measurement accuracy and reproducibility with modularity.
- Measure by suction (samples with high viscosity) and under pressure (liquids with low boiling points)
- Modular design: Up to four ViscoPump II modules
- Multiple measurement station controllable via a PC and multitasking software
- WinVisco 370 software for two rinsing methods
- Virtually all standard capillary viscometers can be used
The software allows control of up to two complete AVS 370 systems or a total of eight ViscoPump II modules. The results can be used and viewed immediately and easily in separate printouts.
Delivery information: The basic version includes a ViscoPump II module for optical detection (for measuring the high or low viscosity of liquids) or a ViscoPump II module with thermocouple temperature detection (for measuring opaque and dark liquids) and WinVisco 370 software in a compact case.
Specifications
- Model:AVS 370 Module 2
- Description:Unit for polymer 2 (230 V)
- Seq:04
- Cat. No.:616-0067
- Measurement range (flow time):Up to 9999,99 seconds
Accessories
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, EPDM (buna), VZ 7051
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, NBR (Perbunan), VZ 7052
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, FKM (viton), VZ 7050
Product Information
- Catalog #AvailabilityPrice
- Safetybottle, without glass thread
Product Information
- Catalog #AvailabilityPrice
- Safety bottle with glass thread, VZ 7021Safety bottle with glass thread, VZ 7021SCOI285420269Log in for availability1 * 1 Items - 1.440,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mmVZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mmSCOI285414048Log in for availability1 * 1 Items - 595,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470SCOI285414031Log in for availability1 * 1 Items - 3.720,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7150 Connection between vacuum pump and both waste bottlesVZ 7150 Connection between vacuum pump and both waste bottlesSCOI285421557Log in for availability1 * 1 Items - 3.720,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mmVZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mmSCOI285410519Log in for availability1 * 1 Items - 443,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mmVZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mmSCOI285410527Log in for availability1 * 1 Items - 443,00 kr
Product Information
- Catalog #AvailabilityPrice
- Fixing frame, stainless steel for micro-OstwaldFixing frame, stainless steel for micro-Ostwald616-3389Log in for availability1 * 1 Items - 4.140,00 kr
Product Information
- Catalog #AvailabilityPrice
- Support PTFE for Cannon-Fenske (routine automatic viscometer)Support PTFE for Cannon-Fenske (routine automatic viscometer)616-3391Log in for availability1 * 1 Items - 2.800,00 kr
Product Information
- Catalog #Temperature RangeMaterialAvailabilityPrice
- Measuring stand AVS/SK-V
Product Information
- Catalog #Temperature RangeAvailabilityPrice
- Measuring stand AVS/S-HT
Product Information
- Catalog #Temperature RangeMaterialAvailabilityPrice
- Measuring stand AVS/SK
Product Information
- Catalog #Temperature RangeAvailabilityPrice
- Measuring stand AVS/S-CF
Product Information
- Catalog #Temperature RangeAvailabilityPrice
- Measuring stand AVS/S
Product Information
- Catalog #AvailabilityPrice
- Fixing frame, stainless steel for Ubbelohde and micro-Ubbelohde viscometersFixing frame, stainless steel for Ubbelohde and micro-Ubbelohde viscometers616-1100Log in for availability1 * 1 Items - 2.210,00 kr
Product Information
- Catalog #AvailabilityPrice
- Absorbing filter with activated coal, VZ 7216Absorbing filter with activated coal, VZ 7216SCOI285422215Log in for availability1 * 1 Items - 1.950,00 kr
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, VZ 7053, 8,3×2,4 mm
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, VZ 7054, 7,0×3,0 mm
Product Information
- Catalog #AvailabilityPrice
- Connectors, VZ 7211, for absorbing filter VZ 7215, 7216Connectors, VZ 7211, for absorbing filter VZ 7215, 7216SCOI285422167Log in for availability1 * 1 Items - 1.020,00 kr
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, VZ 7055, 5,3×1,8 mm
Product Information
- Catalog #AvailabilityPrice
- Absorbing filter, VZ 7215
Product Information
- Catalog #AvailabilityPrice
- Safety device for vacuum line
Product Information
- Catalog #AvailabilityPrice
- VZ 7111 Connecting cable ViscoPump, for TC sensingVZ 7111 Connecting cable ViscoPump, for TC sensingSCOI285421162Log in for availability1 * 1 Items - 1.440,00 kr
Product Information
- Catalog #AvailabilityPrice
- TZ 3855 PTFE piston 50 ml with piston rod for exchangeable unit WA 50 VTZ 3855 PTFE piston 50 ml with piston rod for exchangeable unit WA 50 VSCOI285220700Log in for availability1 * 1 Items - 5.870,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8536 PTFE connection tube, ViscoPump; venting tube viscometerVZ 8536 PTFE connection tube, ViscoPump; venting tube viscometerSCOI1057671Log in for availability1 * 1 Items - 2.960,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5392 Connection piece, PCTFE with M 14 thread (viscometer, capillary connection)VZ 5392 Connection piece, PCTFE with M 14 thread (viscometer, capillary connection)SCOI285411983Log in for availability1 * 1 Items - 1.180,00 kr
Product Information
- Catalog #AvailabilityPrice
- T 500-M1 TITRONIC® 500 basic unit with magnetic stirrer TM 235, with stand rod and titration clamp Z 305, controller TZ 3880, power supply 100 - 240 V; piston burettesT 500-M1 TITRONIC® 500 basic unit with magnetic stirrer TM 235, with stand rod and titration clamp Z 305, controller TZ 3880, power supply 100 - 240 V; piston burettes661-0117Log in for availability1 * 1 Items - 34.700,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5504 Hose set to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 ..,538 .., 547 .., 548 .. and 549 .. to an AVS® 310, 350, 450 and 470VZ 5504 Hose set to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 ..,538 .., 547 .., 548 .. and 549 .. to an AVS® 310, 350, 450 and 470SCOI285414027Log in for availability1 * 1 Items - 3.130,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8551 Balance for waste bottle
Product Information
- Catalog #AvailabilityPrice
- VZ 7116 RS232 data cable, ViscoPump; PC, 'Must be ordered separately'VZ 7116 RS232 data cable, ViscoPump; PC, 'Must be ordered separately'SCOI285421216Log in for availability1 * 1 Items - 2.050,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8610 Connection tube, vacuum pump; waste valve blockVZ 8610 Connection tube, vacuum pump; waste valve blockSCOI1065996Log in for availability1 * 1 Items - 606,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8541 Silicon connecting tube
Product Information
- Catalog #AvailabilityPrice
- VZ 8608 PTFE connection tube, waste bottle top; viscometerVZ 8608 PTFE connection tube, waste bottle top; viscometerSCOI1060765Log in for availability1 * 1 Items - 942,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5391 Connection piece PCTFE with M 10 thread (viscometer, ventilation connection)VZ 5391 Connection piece PCTFE with M 10 thread (viscometer, ventilation connection)SCOI285411991Log in for availability1 * 1 Items - 1.680,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7098 Filter disks for ProClean II filtration system, made of PE, 500 piecesVZ 7098 Filter disks for ProClean II filtration system, made of PE, 500 piecesSCOI285422600Log in for availability1 * 1 Items - 5.340,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5606 Silicon tube/cable set, to connect TC- viscometers with connection threads for viscosity measuring units AVS® 350, 370, 450 und 470; scope of delivery: pressure, ventilation and connecting cable for TC-viscometerVZ 5606 Silicon tube/cable set, to connect TC- viscometers with connection threads for viscosity measuring units AVS® 350, 370, 450 und 470; scope of delivery: pressure, ventilation and connecting cable for TC-viscometerSCOI285413462Log in for availability1 * 1 Items - 3.560,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8634 Filling funnel with connectors for solvent rinsing, used in sample filling set VZ 8604VZ 8634 Filling funnel with connectors for solvent rinsing, used in sample filling set VZ 8604SCOI285422550Log in for availability1 * 1 Items - 2.430,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 6280 PTFE discharge tube, int.Ø 2 mm (viscometer, waste bottle)VZ 6280 PTFE discharge tube, int.Ø 2 mm (viscometer, waste bottle)SCOI285411872Log in for availability1 * 1 Items - 736,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5609 Connection kit for TC viscometer, scope of delivery: tubing set VZ 5606, holding strap VZ 5932 and bracket 053 93VZ 5609 Connection kit for TC viscometer, scope of delivery: tubing set VZ 5606, holding strap VZ 5932 and bracket 053 93SCOI285414759Log in for availability1 * 1 Items - 9.130,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8507 Multilanguage version, including RS cable VZ 7125 to connect to thermostat CT72/52; ViscoSystem® - AVS® 370VZ 8507 Multilanguage version, including RS cable VZ 7125 to connect to thermostat CT72/52; ViscoSystem® - AVS® 370SCOI285424270Log in for availability1 * 1 Items - 22.930,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8540 Silicon connecting tube
Product Information
- Catalog #AvailabilityPrice
- VZ 8603 Filling funnel, packing size: 10 piecesVZ 8603 Filling funnel, packing size: 10 piecesSCOI1064721Log in for availability1 * 10 Items - 1.950,00 kr
Product Information
- Catalog #AvailabilityPrice
- Z 300 Stand foot for the usage without magnetic stirrer TM235, necessary for the titration clamp Z 305Z 300 Stand foot for the usage without magnetic stirrer TM235, necessary for the titration clamp Z 305SCOI285220920Log in for availability1 * 1 Items - 2.220,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8525 PTFE connection tube, int.Ø 2 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4VZ 8525 PTFE connection tube, int.Ø 2 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4SCOI1057673Log in for availability1 * 1 Each - 4.590,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7090 ProClean II, filter system for aggressive samples, contains 100 ml bottles with caps and filter holder (each 5×); filter plates made of Hastelloy C4 (10×20, 10×30 µm)VZ 7090 ProClean II, filter system for aggressive samples, contains 100 ml bottles with caps and filter holder (each 5×); filter plates made of Hastelloy C4 (10×20, 10×30 µm)SCOI285422500Log in for availability1 * 1 Items - 6.290,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5379 Waste bottle, 2000 ml, clear, with NS 29VZ 5379 Waste bottle, 2000 ml, clear, with NS 29SCOI285412363Log in for availability1 * 1 Items - 1.230,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8562 ViscoPump for TC detection including connection cable for TC viscometersVZ 8562 ViscoPump for TC detection including connection cable for TC viscometersSCOI285424070Log in for availability1 * 1 Items - 56.420,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8530 PTFE hose set for high-aggressive media, for Ubbelohde viscometer with discharge tube and thread connection, contains safety bottles with soda limeVZ 8530 PTFE hose set for high-aggressive media, for Ubbelohde viscometer with discharge tube and thread connection, contains safety bottles with soda limeSCOI285415907Log in for availability1 * 1 Items - 11.820,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 6240 Bottle adapter with M10 thread for waste bottle with NS 29 (e.g. VZ 5379)VZ 6240 Bottle adapter with M10 thread for waste bottle with NS 29 (e.g. VZ 5379)SCOI285411486Log in for availability1 * 1 Items - 2.350,00 kr
Product Information
- Catalog #AvailabilityPrice
- T 500 TITRONIC® 500 basic unit without magnetic stirrer, with stand rod and titration clamp Z305, controller TZ 3880, power supply 100 - 240 V; piston burettesT 500 TITRONIC® 500 basic unit without magnetic stirrer, with stand rod and titration clamp Z305, controller TZ 3880, power supply 100 - 240 V; piston burettesSCOI285220200Log in for availability1 * 1 Items - 30.090,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8524 PTFE connection tube, for samples with acid vapour, with sodalime filter, contains: VZ 8537, VZ 8538, VZ 7012, VZ 7211, VZ 7212, VZ 7021, 2×VZ 7215VZ 8524 PTFE connection tube, for samples with acid vapour, with sodalime filter, contains: VZ 8537, VZ 8538, VZ 7012, VZ 7211, VZ 7212, VZ 7021, 2×VZ 7215SCOI1057880Log in for availability1 * 1 Each - 11.800,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5364 Sleeve made of PTFE, reusable
Product Information
- Catalog #AvailabilityPrice
- VZ 8609 PTFE connection tube, waste valve block; waste bottleVZ 8609 PTFE connection tube, waste valve block; waste bottleSCOI1060766Log in for availability1 * 1 Items - 639,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215SCOI285415885Log in for availability1 * 1 Items - 12.310,00 kr
Product Information
- Catalog #AvailabilityPrice
- WA 50 V 50 ml exchangeable burette head for aggresive liquids with integrated chip for reagent data, with brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tipWA 50 V 50 ml exchangeable burette head for aggresive liquids with integrated chip for reagent data, with brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tipSCOI285220360Log in for availability1 * 1 Items - 14.420,00 kr
Product Information
- Catalog #AvailabilityPrice
- TZ 3087, Data cable for connecting TitroLine® alpha plus, TW alpha plus, or Titronic® 110 plus to Titronic® universal, 1,5 mTZ 3087, Data cable for connecting TitroLine® alpha plus, TW alpha plus, or Titronic® 110 plus to Titronic® universal, 1,5 mSCOI1007976Log in for availability1 * 1,5 m - 1.210,00 kr
Product Information
- Catalog #AvailabilityPrice
- TZ 3425 Hose made out of FEP, Ø3×0,5 mm, 5 m lengthTZ 3425 Hose made out of FEP, Ø3×0,5 mm, 5 m lengthSCOI285216020Log in for availability1 * 1 Items - 747,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5525 Union nut, PPS, with M 14 thread for VZ 5392VZ 5525 Union nut, PPS, with M 14 thread for VZ 5392SCOI285411918Log in for availability1 * 1 Items - 286,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8602Valve block includes mounting devices and 4 solenoid valvesVZ 8602Valve block includes mounting devices and 4 solenoid valvesSCOI1063704Log in for availability1 * 1 Items - 35.480,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7097 Filter disks for ProClean II filtration system, made of PE, 50 piecesVZ 7097 Filter disks for ProClean II filtration system, made of PE, 50 piecesSCOI285422590Log in for availability1 * 1 Items - 769,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8502 Multiple power version (95 - 230 V) for thermal detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370VZ 8502 Multiple power version (95 - 230 V) for thermal detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370SCOI1062837Log in for availability1 * 1 Items - 123.790,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7096 Bottle set for ProClean II, scope of delivery: 100 ml Flaschen with screw caps (with and without borehole, 5 pieces each)VZ 7096 Bottle set for ProClean II, scope of delivery: 100 ml Flaschen with screw caps (with and without borehole, 5 pieces each)SCOI285422490Log in for availability1 * 1 Items - 1.230,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8561 ViscoPump for optical detection, scope of delivery: connection cable for measuring stand, tube holder VZ 5127 and holder for safety bottleVZ 8561 ViscoPump for optical detection, scope of delivery: connection cable for measuring stand, tube holder VZ 5127 and holder for safety bottleSCOI285424060Log in for availability1 * 1 Items - 51.540,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8501 Multiple power version (95 - 230 V) for optical detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece, ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370VZ 8501 Multiple power version (95 - 230 V) for optical detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece, ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370SCOI1062836Log in for availability1 * 1 Items - 123.790,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8522 PTFE connection tube, int.Ø 2 mm, short tubings (40 cm length) for AVS®370 - alternative to long tubings VZ 5504, only suitable, if AVS370 is positioned on instrument support VZ 8571VZ 8522 PTFE connection tube, int.Ø 2 mm, short tubings (40 cm length) for AVS®370 - alternative to long tubings VZ 5504, only suitable, if AVS370 is positioned on instrument support VZ 8571SCOI1058420Log in for availability1 * 1 Each - 2.820,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8622 PTFE connection tube, sample filling funnel; viscometerVZ 8622 PTFE connection tube, sample filling funnel; viscometerSCOI1058398Log in for availability1 * 1 Items - 1.410,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8526 Upgrading set for viscopump to vacuum mode in addition to VZ 5505VZ 8526 Upgrading set for viscopump to vacuum mode in addition to VZ 5505SCOI285420530Log in for availability1 * 1 Items - 2.900,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8606 waste basic set, contains: Tubes VZ 8608 and VZ 8609, 2 ml waste bottle, bottle top VZ 6240 and PTFE cuff NS 29VZ 8606 waste basic set, contains: Tubes VZ 8608 and VZ 8609, 2 ml waste bottle, bottle top VZ 6240 and PTFE cuff NS 29SCOI1063833Log in for availability1 * 1 Items - 5.380,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7094 Filter blanks for ProClean II, Hastelloy C4, pore size 20 µm, 10 piecesVZ 7094 Filter blanks for ProClean II, Hastelloy C4, pore size 20 µm, 10 piecesSCOI285422480Log in for availability1 * 1 Items - 1.940,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 6286 PTFE discharge tube, int.Ø 1,5 mm (viscometer, waste bottle)VZ 6286 PTFE discharge tube, int.Ø 1,5 mm (viscometer, waste bottle)SCOI285411729Log in for availability1 * 1 Items - 909,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7093 Screw caps for filter, ProClean II, 10 piecesVZ 7093 Screw caps for filter, ProClean II, 10 piecesSCOI285422510Log in for availability1 * 1 Items - 563,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8539 Silicon connecting tube
Product Information
- Catalog #AvailabilityPrice
- VZ 8630 Vacuum pump, version 230 V, 50 Hz For AVS® 370 rinsing and waste system and AVS® ProVZ 8630 Vacuum pump, version 230 V, 50 Hz For AVS® 370 rinsing and waste system and AVS® ProSCOI1057901Log in for availability1 * 1 Items - 43.650,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8604 Sample filling set, contains: funnel with 2 rinsing connections, filling tube, funnel VZ 8603, sealing cap VZ 8605 and adapterVZ 8604 Sample filling set, contains: funnel with 2 rinsing connections, filling tube, funnel VZ 8603, sealing cap VZ 8605 and adapterSCOI1061389Log in for availability1 * 1 Each - 5.080,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523SCOI1057903Log in for availability1 * 1 Items - 5.840,00 kr
Product Information
- Catalog #AvailabilityPrice
- TZ 3886 Titration stand without stirring function, for TZ 3885 as option for magnetic stirrer TM 50TZ 3886 Titration stand without stirring function, for TZ 3885 as option for magnetic stirrer TM 50SCOI285225910Log in for availability1 * 1 Items - 920,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8523 PTFE connection tube, int.Ø 1,5 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4VZ 8523 PTFE connection tube, int.Ø 1,5 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4SCOI1057648Log in for availability1 * 1 Each - 6.250,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5932 Holder for TC-Viscometer, including mounting devices for filling funnel and safety bottleVZ 5932 Holder for TC-Viscometer, including mounting devices for filling funnel and safety bottleSCOI285413865Log in for availability1 * 1 Items - 7.750,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7095 Filter blanks for ProClean II, Hastelloy C4, pore size 30 µm, 10 piecesVZ 7095 Filter blanks for ProClean II, Hastelloy C4, pore size 30 µm, 10 piecesSCOI285422520Log in for availability1 * 1 Items - 1.940,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8535 PTFE connection tube, ViscoPump; safety bottle VZ 7021, for AVS®370VZ 8535 PTFE connection tube, ViscoPump; safety bottle VZ 7021, for AVS®370SCOI1057672Log in for availability1 * 1 Items - 1.260,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7115 RS232 data cable ViscoPump; ViscoPump, 'Must be ordered separately'VZ 7115 RS232 data cable ViscoPump; ViscoPump, 'Must be ordered separately'SCOI285421208Log in for availability1 * 1 Items - 1.440,00 kr
Product Information
- Catalog #AvailabilityPrice
- TZ 1512 Dosage tip, 120 mm length, capillary Ø 1,26 mm, for WA 50 V; titration tipsTZ 1512 Dosage tip, 120 mm length, capillary Ø 1,26 mm, for WA 50 V; titration tipsSCOI285213738Log in for availability1 * 1 Items - 725,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 5524 Union nut, PPS, with M 10 thread for VZ 5391VZ 5524 Union nut, PPS, with M 10 thread for VZ 5391SCOI285411926Log in for availability1 * 1 Items - 286,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7110 Connecting cable ViscoPump, for optical sensingVZ 7110 Connecting cable ViscoPump, for optical sensingSCOI285421154Log in for availability1 * 1 Items - 1.270,00 kr
Product Information
- Catalog #AvailabilityPrice
- TZ 3094 Data cable RS-232, 1st + 2nd side: 4-pole plug mini DIN, connection: TITRONIC® universal/500 new titrators; TITRONIC® universal/500, all new titrators, 1,0 mTZ 3094 Data cable RS-232, 1st + 2nd side: 4-pole plug mini DIN, connection: TITRONIC® universal/500 new titrators; TITRONIC® universal/500, all new titrators, 1,0 mSCOI285223545Log in for availability1 * 1 Items - 628,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 8601 Valve block includes mounting devices and 2 solenoid valvesVZ 8601 Valve block includes mounting devices and 2 solenoid valvesSCOI1063702Log in for availability1 * 1 Items - 18.850,00 kr
Product Information
- Catalog #AvailabilityPrice
- VZ 7092 Filter holder for ProClean II, 5 piecesVZ 7092 Filter holder for ProClean II, 5 piecesSCOI285422470Log in for availability1 * 1 Items - 2.740,00 kr