Order Entry
Denmark
Orders LinkContactUsLinkComponent
Automatic capillary viscometers, ViscoSystem® AVC 370
Unit for polymer 1 (230 V), AVS 370 Module 1
Catalog # SCOI1056516
Supplier:   SI Analytics
Automatic capillary viscometers, ViscoSystem® AVC 370
Catalog # SCOI1056516
Supplier:  SI Analytics
Automatic capillary viscometers, ViscoSystem® AVC 370
Automatic capillary viscometers, ViscoSystem® AVC 370
Unit for polymer 1 (230 V), AVS 370 Module 1
Price
kr345,820.00
Log in for availability
1 * 1 Items
Product Details & Documents
ViscoSystem AVC 370 combines a high degree of measurement accuracy and reproducibility with modularity.
  • Measure by suction (samples with high viscosity) and under pressure (liquids with low boiling points)
  • Modular design: Up to four ViscoPump II modules
  • Multiple measurement station controllable via a PC and multitasking software
  • WinVisco 370 software for two rinsing methods
  • Virtually all standard capillary viscometers can be used
The software allows control of up to two complete AVS 370 systems or a total of eight ViscoPump II modules. The results can be used and viewed immediately and easily in separate printouts.
 : The basic version includes a ViscoPump II module for optical detection (for measuring the high or low viscosity of liquids) or a ViscoPump II module with thermocouple temperature detection (for measuring opaque and dark liquids) and WinVisco 370 software in a compact case.
Specifications
  • Model:
    AVS 370 Module 1
  • Description:
    Unit for polymer 1 (230 V)
  • Seq:
    03
  • Cat. No.:
    SCOI1056516
  • Measurement range (flow time):
    Up to 9999,99 seconds
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, EPDM (buna), VZ 7051
    O-ring seal, EPDM (buna), VZ 7051
    SCOI285420569
    Log in for availability
    1 * 1 Items - 248,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, NBR (Perbunan), VZ 7052
    O-ring seal, NBR (Perbunan), VZ 7052
    SCOI285420577
    Log in for availability
    1 * 1 Items - 235,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, FKM (viton), VZ 7050
    O-ring seal, FKM (viton), VZ 7050
    SCOI285420552
    Log in for availability
    1 * 1 Items - 248,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Safetybottle, without glass thread
    Safetybottle, without glass thread
    SCOI285420277
    Log in for availability
    1 * 1 Items - 1.180,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Safety bottle with glass thread, VZ 7021
    Safety bottle with glass thread, VZ 7021
    SCOI285420269
    Log in for availability
    1 * 1 Items - 1.440,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mm
    VZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mm
    SCOI285414048
    Log in for availability
    1 * 1 Items - 595,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470
    VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470
    SCOI285414031
    Log in for availability
    1 * 1 Items - 3.720,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7150 Connection between vacuum pump and both waste bottles
    VZ 7150 Connection between vacuum pump and both waste bottles
    SCOI285421557
    Log in for availability
    1 * 1 Items - 3.720,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mm
    VZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mm
    SCOI285410519
    Log in for availability
    1 * 1 Items - 443,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mm
    VZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mm
    SCOI285410527
    Log in for availability
    1 * 1 Items - 443,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Fixing frame, stainless steel for micro-Ostwald
    Fixing frame, stainless steel for micro-Ostwald
    616-3389
    Log in for availability
    1 * 1 Items - 4.140,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Support PTFE for Cannon-Fenske (routine automatic viscometer)
    Support PTFE for Cannon-Fenske (routine automatic viscometer)
    616-3391
    Log in for availability
    1 * 1 Items - 2.800,00 kr
Product Information
  • Catalog #
    Temperature Range
    Material
    Availability
    Price
  • Measuring stand AVS/SK-V
    Measuring stand AVS/SK-V
    616-3396
    0...60 °C
    PVDF
    Log in for availability
    1 * 1 Items - 43.710,00 kr
Product Information
  • Catalog #
    Temperature Range
    Availability
    Price
  • Measuring stand AVS/S-HT
    Measuring stand AVS/S-HT
    616-3398
    -80...+200 °C
    Log in for availability
    1 * 1 Items - 43.650,00 kr
Product Information
  • Catalog #
    Temperature Range
    Material
    Availability
    Price
  • Measuring stand AVS/SK
    Measuring stand AVS/SK
    616-3394
    0...60 °C
    PVDF
    Log in for availability
    1 * 1 Items - 39.010,00 kr
Product Information
  • Catalog #
    Temperature Range
    Availability
    Price
  • Measuring stand AVS/S-CF
    Measuring stand AVS/S-CF
    616-3395
    -80...+100 °C
    Log in for availability
    1 * 1 Items - 43.080,00 kr
Product Information
  • Catalog #
    Temperature Range
    Availability
    Price
  • Measuring stand AVS/S
    Measuring stand AVS/S
    616-3393
    -80...+100 °C
    Log in for availability
    1 * 1 Items - 37.020,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Fixing frame, stainless steel for Ubbelohde and micro-Ubbelohde viscometers
    Fixing frame, stainless steel for Ubbelohde and micro-Ubbelohde viscometers
    616-1100
    Log in for availability
    1 * 1 Items - 2.210,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Absorbing filter with activated coal, VZ 7216
    Absorbing filter with activated coal, VZ 7216
    SCOI285422215
    Log in for availability
    1 * 1 Items - 1.950,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, VZ 7053, 8,3×2,4 mm
    O-ring seal, VZ 7053, 8,3×2,4 mm
    SCOI285420585
    Log in for availability
    1 * 1 Items - 2.050,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, VZ 7054, 7,0×3,0 mm
    O-ring seal, VZ 7054, 7,0×3,0 mm
    SCOI285420593
    Log in for availability
    1 * 1 Items - 1.750,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Connectors, VZ 7211, for absorbing filter VZ 7215, 7216
    Connectors, VZ 7211, for absorbing filter VZ 7215, 7216
    SCOI285422167
    Log in for availability
    1 * 1 Items - 1.020,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, VZ 7055, 5,3×1,8 mm
    O-ring seal, VZ 7055, 5,3×1,8 mm
    SCOI285420606
    Log in for availability
    1 * 1 Items - 1.270,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Absorbing filter, VZ 7215
    Absorbing filter, VZ 7215
    SCOI285422204
    Log in for availability
    1 * 1 Items - 1.960,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Safety device for vacuum line
    Safety device for vacuum line
    SCOI1054303
    Log in for availability
    1 * 1 Items - 5.480,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7111 Connecting cable ViscoPump, for TC sensing
    VZ 7111 Connecting cable ViscoPump, for TC sensing
    SCOI285421162
    Log in for availability
    1 * 1 Items - 1.440,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • TZ 3855 PTFE piston 50 ml with piston rod for exchangeable unit WA 50 V
    TZ 3855 PTFE piston 50 ml with piston rod for exchangeable unit WA 50 V
    SCOI285220700
    Log in for availability
    1 * 1 Items - 5.870,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8536 PTFE connection tube, ViscoPump; venting tube viscometer
    VZ 8536 PTFE connection tube, ViscoPump; venting tube viscometer
    SCOI1057671
    Log in for availability
    1 * 1 Items - 2.960,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5392 Connection piece, PCTFE with M 14 thread (viscometer, capillary connection)
    VZ 5392 Connection piece, PCTFE with M 14 thread (viscometer, capillary connection)
    SCOI285411983
    Log in for availability
    1 * 1 Items - 1.180,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • T 500-M1 TITRONIC® 500 basic unit with magnetic stirrer TM 235, with stand rod and titration clamp Z 305, controller TZ 3880, power supply 100 - 240 V; piston burettes
    T 500-M1 TITRONIC® 500 basic unit with magnetic stirrer TM 235, with stand rod and titration clamp Z 305, controller TZ 3880, power supply 100 - 240 V; piston burettes
    661-0117
    Log in for availability
    1 * 1 Items - 34.700,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5504 Hose set to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 ..,538 .., 547 .., 548 .. and 549 .. to an AVS® 310, 350, 450 and 470
    VZ 5504 Hose set to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 ..,538 .., 547 .., 548 .. and 549 .. to an AVS® 310, 350, 450 and 470
    SCOI285414027
    Log in for availability
    1 * 1 Items - 3.130,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8551 Balance for waste bottle
    VZ 8551 Balance for waste bottle
    SCOI1054112
    Log in for availability
    1 * 1 Items - 15.220,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7116 RS232 data cable, ViscoPump; PC, 'Must be ordered separately'
    VZ 7116 RS232 data cable, ViscoPump; PC, 'Must be ordered separately'
    SCOI285421216
    Log in for availability
    1 * 1 Items - 2.050,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8610 Connection tube, vacuum pump; waste valve block
    VZ 8610 Connection tube, vacuum pump; waste valve block
    SCOI1065996
    Log in for availability
    1 * 1 Items - 606,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8541 Silicon connecting tube
    VZ 8541 Silicon connecting tube
    SCOI1057676
    Log in for availability
    1 * 1 Items - 855,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8608 PTFE connection tube, waste bottle top; viscometer
    VZ 8608 PTFE connection tube, waste bottle top; viscometer
    SCOI1060765
    Log in for availability
    1 * 1 Items - 942,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5391 Connection piece PCTFE with M 10 thread (viscometer, ventilation connection)
    VZ 5391 Connection piece PCTFE with M 10 thread (viscometer, ventilation connection)
    SCOI285411991
    Log in for availability
    1 * 1 Items - 1.680,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7098 Filter disks for ProClean II filtration system, made of PE, 500 pieces
    VZ 7098 Filter disks for ProClean II filtration system, made of PE, 500 pieces
    SCOI285422600
    Log in for availability
    1 * 1 Items - 5.340,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5606 Silicon tube/cable set, to connect TC- viscometers with connection threads for viscosity measuring units AVS® 350, 370, 450 und 470; scope of delivery: pressure, ventilation and connecting cable for TC-viscometer
    VZ 5606 Silicon tube/cable set, to connect TC- viscometers with connection threads for viscosity measuring units AVS® 350, 370, 450 und 470; scope of delivery: pressure, ventilation and connecting cable for TC-viscometer
    SCOI285413462
    Log in for availability
    1 * 1 Items - 3.560,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8634 Filling funnel with connectors for solvent rinsing, used in sample filling set VZ 8604
    VZ 8634 Filling funnel with connectors for solvent rinsing, used in sample filling set VZ 8604
    SCOI285422550
    Log in for availability
    1 * 1 Items - 2.430,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 6280 PTFE discharge tube, int.Ø 2 mm (viscometer, waste bottle)
    VZ 6280 PTFE discharge tube, int.Ø 2 mm (viscometer, waste bottle)
    SCOI285411872
    Log in for availability
    1 * 1 Items - 736,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5609 Connection kit for TC viscometer, scope of delivery: tubing set VZ 5606, holding strap VZ 5932 and bracket 053 93
    VZ 5609 Connection kit for TC viscometer, scope of delivery: tubing set VZ 5606, holding strap VZ 5932 and bracket 053 93
    SCOI285414759
    Log in for availability
    1 * 1 Items - 9.130,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8507 Multilanguage version, including RS cable VZ 7125 to connect to thermostat CT72/52; ViscoSystem® - AVS® 370
    VZ 8507 Multilanguage version, including RS cable VZ 7125 to connect to thermostat CT72/52; ViscoSystem® - AVS® 370
    SCOI285424270
    Log in for availability
    1 * 1 Items - 22.930,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8540 Silicon connecting tube
    VZ 8540 Silicon connecting tube
    SCOI1057675
    Log in for availability
    1 * 1 Items - 887,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8603 Filling funnel, packing size: 10 pieces
    VZ 8603 Filling funnel, packing size: 10 pieces
    SCOI1064721
    Log in for availability
    1 * 10 Items - 1.950,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Z 300 Stand foot for the usage without magnetic stirrer TM235, necessary for the titration clamp Z 305
    Z 300 Stand foot for the usage without magnetic stirrer TM235, necessary for the titration clamp Z 305
    SCOI285220920
    Log in for availability
    1 * 1 Items - 2.220,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8525 PTFE connection tube, int.Ø 2 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4
    VZ 8525 PTFE connection tube, int.Ø 2 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4
    SCOI1057673
    Log in for availability
    1 * 1 Each - 4.590,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7090 ProClean II, filter system for aggressive samples, contains 100 ml bottles with caps and filter holder (each 5×); filter plates made of Hastelloy C4 (10×20, 10×30 µm)
    VZ 7090 ProClean II, filter system for aggressive samples, contains 100 ml bottles with caps and filter holder (each 5×); filter plates made of Hastelloy C4 (10×20, 10×30 µm)
    SCOI285422500
    Log in for availability
    1 * 1 Items - 6.290,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5379 Waste bottle, 2000 ml, clear, with NS 29
    VZ 5379 Waste bottle, 2000 ml, clear, with NS 29
    SCOI285412363
    Log in for availability
    1 * 1 Items - 1.230,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8562 ViscoPump for TC detection including connection cable for TC viscometers
    VZ 8562 ViscoPump for TC detection including connection cable for TC viscometers
    SCOI285424070
    Log in for availability
    1 * 1 Items - 56.420,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8530 PTFE hose set for high-aggressive media, for Ubbelohde viscometer with discharge tube and thread connection, contains safety bottles with soda lime
    VZ 8530 PTFE hose set for high-aggressive media, for Ubbelohde viscometer with discharge tube and thread connection, contains safety bottles with soda lime
    SCOI285415907
    Log in for availability
    1 * 1 Items - 11.820,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 6240 Bottle adapter with M10 thread for waste bottle with NS 29 (e.g. VZ 5379)
    VZ 6240 Bottle adapter with M10 thread for waste bottle with NS 29 (e.g. VZ 5379)
    SCOI285411486
    Log in for availability
    1 * 1 Items - 2.350,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • T 500 TITRONIC® 500 basic unit without magnetic stirrer, with stand rod and titration clamp Z305, controller TZ 3880, power supply 100 - 240 V; piston burettes
    T 500 TITRONIC® 500 basic unit without magnetic stirrer, with stand rod and titration clamp Z305, controller TZ 3880, power supply 100 - 240 V; piston burettes
    SCOI285220200
    Log in for availability
    1 * 1 Items - 30.090,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8524 PTFE connection tube, for samples with acid vapour, with sodalime filter, contains: VZ 8537, VZ 8538, VZ 7012, VZ 7211, VZ 7212, VZ 7021, 2×VZ 7215
    VZ 8524 PTFE connection tube, for samples with acid vapour, with sodalime filter, contains: VZ 8537, VZ 8538, VZ 7012, VZ 7211, VZ 7212, VZ 7021, 2×VZ 7215
    SCOI1057880
    Log in for availability
    1 * 1 Each - 11.800,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5364 Sleeve made of PTFE, reusable
    VZ 5364 Sleeve made of PTFE, reusable
    SCOI285412417
    Log in for availability
    1 * 1 Items - 181,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8609 PTFE connection tube, waste valve block; waste bottle
    VZ 8609 PTFE connection tube, waste valve block; waste bottle
    SCOI1060766
    Log in for availability
    1 * 1 Items - 639,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215
    VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215
    SCOI285415885
    Log in for availability
    1 * 1 Items - 12.310,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • WA 50 V 50 ml exchangeable burette head for aggresive liquids with integrated chip for reagent data, with brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tip
    WA 50 V 50 ml exchangeable burette head for aggresive liquids with integrated chip for reagent data, with brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tip
    SCOI285220360
    Log in for availability
    1 * 1 Items - 14.420,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • TZ 3087, Data cable for connecting TitroLine® alpha plus, TW alpha plus, or Titronic® 110 plus to Titronic® universal, 1,5 m
    TZ 3087, Data cable for connecting TitroLine® alpha plus, TW alpha plus, or Titronic® 110 plus to Titronic® universal, 1,5 m
    SCOI1007976
    Log in for availability
    1 * 1,5 m - 1.210,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • TZ 3425 Hose made out of FEP, Ø3×0,5 mm, 5 m length
    TZ 3425 Hose made out of FEP, Ø3×0,5 mm, 5 m length
    SCOI285216020
    Log in for availability
    1 * 1 Items - 747,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5525 Union nut, PPS, with M 14 thread for VZ 5392
    VZ 5525 Union nut, PPS, with M 14 thread for VZ 5392
    SCOI285411918
    Log in for availability
    1 * 1 Items - 286,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8602Valve block includes mounting devices and 4 solenoid valves
    VZ 8602Valve block includes mounting devices and 4 solenoid valves
    SCOI1063704
    Log in for availability
    1 * 1 Items - 35.480,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7097 Filter disks for ProClean II filtration system, made of PE, 50 pieces
    VZ 7097 Filter disks for ProClean II filtration system, made of PE, 50 pieces
    SCOI285422590
    Log in for availability
    1 * 1 Items - 769,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8502 Multiple power version (95 - 230 V) for thermal detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370
    VZ 8502 Multiple power version (95 - 230 V) for thermal detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370
    SCOI1062837
    Log in for availability
    1 * 1 Items - 123.790,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7096 Bottle set for ProClean II, scope of delivery: 100 ml Flaschen with screw caps (with and without borehole, 5 pieces each)
    VZ 7096 Bottle set for ProClean II, scope of delivery: 100 ml Flaschen with screw caps (with and without borehole, 5 pieces each)
    SCOI285422490
    Log in for availability
    1 * 1 Items - 1.230,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8561 ViscoPump for optical detection, scope of delivery: connection cable for measuring stand, tube holder VZ 5127 and holder for safety bottle
    VZ 8561 ViscoPump for optical detection, scope of delivery: connection cable for measuring stand, tube holder VZ 5127 and holder for safety bottle
    SCOI285424060
    Log in for availability
    1 * 1 Items - 51.540,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8501 Multiple power version (95 - 230 V) for optical detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece, ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370
    VZ 8501 Multiple power version (95 - 230 V) for optical detection of measuring position 5 - 8, scope of delivery: AVS® 370 basic unit, 1 piece, ViscoPump III, datacable VZ 7115; ViscoSystem® - AVS® 370
    SCOI1062836
    Log in for availability
    1 * 1 Items - 123.790,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8522 PTFE connection tube, int.Ø 2 mm, short tubings (40 cm length) for AVS®370 - alternative to long tubings VZ 5504, only suitable, if AVS370 is positioned on instrument support VZ 8571
    VZ 8522 PTFE connection tube, int.Ø 2 mm, short tubings (40 cm length) for AVS®370 - alternative to long tubings VZ 5504, only suitable, if AVS370 is positioned on instrument support VZ 8571
    SCOI1058420
    Log in for availability
    1 * 1 Each - 2.820,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8622 PTFE connection tube, sample filling funnel; viscometer
    VZ 8622 PTFE connection tube, sample filling funnel; viscometer
    SCOI1058398
    Log in for availability
    1 * 1 Items - 1.410,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8526 Upgrading set for viscopump to vacuum mode in addition to VZ 5505
    VZ 8526 Upgrading set for viscopump to vacuum mode in addition to VZ 5505
    SCOI285420530
    Log in for availability
    1 * 1 Items - 2.900,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8606 waste basic set, contains: Tubes VZ 8608 and VZ 8609, 2 ml waste bottle, bottle top VZ 6240 and PTFE cuff NS 29
    VZ 8606 waste basic set, contains: Tubes VZ 8608 and VZ 8609, 2 ml waste bottle, bottle top VZ 6240 and PTFE cuff NS 29
    SCOI1063833
    Log in for availability
    1 * 1 Items - 5.380,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7094 Filter blanks for ProClean II, Hastelloy C4, pore size 20 µm, 10 pieces
    VZ 7094 Filter blanks for ProClean II, Hastelloy C4, pore size 20 µm, 10 pieces
    SCOI285422480
    Log in for availability
    1 * 1 Items - 1.940,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 6286 PTFE discharge tube, int.Ø 1,5 mm (viscometer, waste bottle)
    VZ 6286 PTFE discharge tube, int.Ø 1,5 mm (viscometer, waste bottle)
    SCOI285411729
    Log in for availability
    1 * 1 Items - 909,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7093 Screw caps for filter, ProClean II, 10 pieces
    VZ 7093 Screw caps for filter, ProClean II, 10 pieces
    SCOI285422510
    Log in for availability
    1 * 1 Items - 563,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8539 Silicon connecting tube
    VZ 8539 Silicon connecting tube
    SCOI1057674
    Log in for availability
    1 * 1 Items - 1.210,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8630 Vacuum pump, version 230 V, 50 Hz For AVS® 370 rinsing and waste system and AVS® Pro
    VZ 8630 Vacuum pump, version 230 V, 50 Hz For AVS® 370 rinsing and waste system and AVS® Pro
    SCOI1057901
    Log in for availability
    1 * 1 Items - 43.650,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8604 Sample filling set, contains: funnel with 2 rinsing connections, filling tube, funnel VZ 8603, sealing cap VZ 8605 and adapter
    VZ 8604 Sample filling set, contains: funnel with 2 rinsing connections, filling tube, funnel VZ 8603, sealing cap VZ 8605 and adapter
    SCOI1061389
    Log in for availability
    1 * 1 Each - 5.080,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523
    VZ 8571 Instrument support for CT 72/2 and CT 72/4, required for measuring systems AVS®370 in case of short hoses between ViscoPump and viscometer, e.g. with hose set VZ 8523
    SCOI1057903
    Log in for availability
    1 * 1 Items - 5.840,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • TZ 3886 Titration stand without stirring function, for TZ 3885 as option for magnetic stirrer TM 50
    TZ 3886 Titration stand without stirring function, for TZ 3885 as option for magnetic stirrer TM 50
    SCOI285225910
    Log in for availability
    1 * 1 Items - 920,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8523 PTFE connection tube, int.Ø 1,5 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4
    VZ 8523 PTFE connection tube, int.Ø 1,5 mm, suitable to thermostat type: CT 53, CT 54, CT 72/2 and CT 72/4
    SCOI1057648
    Log in for availability
    1 * 1 Each - 6.250,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5932 Holder for TC-Viscometer, including mounting devices for filling funnel and safety bottle
    VZ 5932 Holder for TC-Viscometer, including mounting devices for filling funnel and safety bottle
    SCOI285413865
    Log in for availability
    1 * 1 Items - 7.750,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7095 Filter blanks for ProClean II, Hastelloy C4, pore size 30 µm, 10 pieces
    VZ 7095 Filter blanks for ProClean II, Hastelloy C4, pore size 30 µm, 10 pieces
    SCOI285422520
    Log in for availability
    1 * 1 Items - 1.940,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8535 PTFE connection tube, ViscoPump; safety bottle VZ 7021, for AVS®370
    VZ 8535 PTFE connection tube, ViscoPump; safety bottle VZ 7021, for AVS®370
    SCOI1057672
    Log in for availability
    1 * 1 Items - 1.260,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7115 RS232 data cable ViscoPump; ViscoPump, 'Must be ordered separately'
    VZ 7115 RS232 data cable ViscoPump; ViscoPump, 'Must be ordered separately'
    SCOI285421208
    Log in for availability
    1 * 1 Items - 1.440,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • TZ 1512 Dosage tip, 120 mm length, capillary Ø 1,26 mm, for WA 50 V; titration tips
    TZ 1512 Dosage tip, 120 mm length, capillary Ø 1,26 mm, for WA 50 V; titration tips
    SCOI285213738
    Log in for availability
    1 * 1 Items - 725,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5524 Union nut, PPS, with M 10 thread for VZ 5391
    VZ 5524 Union nut, PPS, with M 10 thread for VZ 5391
    SCOI285411926
    Log in for availability
    1 * 1 Items - 286,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7110 Connecting cable ViscoPump, for optical sensing
    VZ 7110 Connecting cable ViscoPump, for optical sensing
    SCOI285421154
    Log in for availability
    1 * 1 Items - 1.270,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • TZ 3094 Data cable RS-232, 1st + 2nd side: 4-pole plug mini DIN, connection: TITRONIC® universal/500 new titrators; TITRONIC® universal/500, all new titrators, 1,0 m
    TZ 3094 Data cable RS-232, 1st + 2nd side: 4-pole plug mini DIN, connection: TITRONIC® universal/500 new titrators; TITRONIC® universal/500, all new titrators, 1,0 m
    SCOI285223545
    Log in for availability
    1 * 1 Items - 628,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 8601 Valve block includes mounting devices and 2 solenoid valves
    VZ 8601 Valve block includes mounting devices and 2 solenoid valves
    SCOI1063702
    Log in for availability
    1 * 1 Items - 18.850,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • VZ 7092 Filter holder for ProClean II, 5 pieces
    VZ 7092 Filter holder for ProClean II, 5 pieces
    SCOI285422470
    Log in for availability
    1 * 1 Items - 2.740,00 kr