Order Entry
Denmark
Orders LinkContactUsLinkComponent
Temperature probes

Temperature probes

Sensor, M16×1, for flow or return double, −90…+200 °C, 1,5 m, - mm
Catalog # HUBE6353
Supplier:   HUBER
Temperature probes
Catalog # HUBE6353
Supplier:  HUBER
Temperature probes
Temperature probes

Sensor, M16×1, for flow or return double, −90…+200 °C, 1,5 m, - mm

Price
5.030,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

For external thermo regulation applications.

Specifications
  • Range:
    −90…+200 °C
  • Description:
    Sensor, M16×1, for flow or return double
  • Length:
    1,5 m
  • Seq:
    08
  • Ø×L:
    - mm