Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Temperature probes

Sensor, M16×1, for flow or return, −90…+200 °C, 1,5 m, - mm
  HUBE6352
 :   HUBER
Temperature probes
  HUBE6352
 :  HUBER
Temperature probes
Temperature probes

Sensor, M16×1, for flow or return, −90…+200 °C, 1,5 m, - mm

 
4.530,00 kr
   
1 * 1 Items
 

For external thermo regulation applications.

 
  • Range:
    −90…+200 °C
  • Description:
    Sensor, M16×1, for flow or return
  • Length:
    1,5 m
  • Seq:
    07
  • Ø×L:
    - mm