Order Entry
Denmark
Orders LinkContactUsLinkComponent
Temperature probes

Temperature probes

Extension cable, PT100, −90…+200 °C, 3 m, - mm
Catalog # HUBE6292
Supplier:   HUBER
Temperature probes
Catalog # HUBE6292
Supplier:  HUBER
Temperature probes
Temperature probes

Extension cable, PT100, −90…+200 °C, 3 m, - mm

Price
2.230,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

For external thermo regulation applications.

Specifications
  • Range:
    −90…+200 °C
  • Description:
    Extension cable, PT100
  • Length:
    3 m
  • Seq:
    06
  • Ø×L:
    - mm