Order Entry
Denmark
Orders LinkContactUsLinkComponent
Scoops, EasyScoop

Scoops, EasyScoop

2000 ml, 300 mm
Catalog # BURK5356-2000
Supplier:   BURKLE
Scoops, EasyScoop
Catalog # BURK5356-2000
Supplier:  BURKLE
Scoops, EasyScoop
Scoops, EasyScoop

2000 ml, 300 mm

Price
515,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Sampling scoops made of light aluminium, bright-anodised.

  • Ergonomically designed: With centrally arranged inner grip
  • Considerably shorter than conventional scoops: Easier to take samples from narrow containers
Specifications
  • Capacity:
    2000 ml
  • Length:
    300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.