WHEATON® Rubber Stoppers
Stopper, grey chlorobutyl, 13,0 mm, 20,0 mm
Catalog # 215-4717
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Stoppers of various styles, sizes and rubber formulations.
- Manufactured from the highest quality raw materials
- Components manufactured to yield exceptional dimensional stability
- Autoclavable
Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.
Snap-on style provides maximum sealing for vials with a blow-back glass finish.
Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.
Documents
Specifications
- Ø bottom:13,0 mm
- Ø top:20,0 mm
- Description:Stopper, grey chlorobutyl
- Seq:19
- Cat. No.:215-4717
- Durometer hardness:46



