Order Entry
Denmark
Orders LinkContactUsLinkComponent
Multidishes, Nunc™ UpCell™ surface
Multidishes, Nunc™ UpCell™ surface
6-well Multidish, UpCell™, vented, with lid, membrane included, 9,6 cm², 3,0 ml
Catalog # 734-2252
Multidishes, Nunc™ UpCell™ surface
Catalog # 734-2252
Multidishes, Nunc™ UpCell™ surface
Multidishes, Nunc™ UpCell™ surface
6-well Multidish, UpCell™, vented, with lid, membrane included, 9,6 cm², 3,0 ml
Price
1.970,00 kr
Log in for availability
1 * 6 Items
Product Details & Documents
The UpCell™ surface is a temperature-responsive cell culture surface that enables harvesting of cells with high viability and intact surface proteins for culture passaging, single-cell analyses and cell transplantation research. The UpCell™ surface enables harvesting of cell sheets and creation of 3D tissue models held together by normal cell junctions and extracellular matrix deposited by the cells.
  • No trypsinisation - cell surface proteins are preserved
  • High cell viability without scraping
  • Releases adherent cells by reduction of temperature of the cell culture
  • Requires minimal hands-on time
Specifications
  • Recommended working volume:
    3,0 ml
  • Description:
    6-well Multidish, UpCell™, vented, with lid, membrane included
  • Seq:
    01
  • Culture area:
    9,6 cm²
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Membrane supports for Nunc™ 12-well multidishes with UpCell™ surface
    Membrane supports for Nunc™ 12-well multidishes with UpCell™ surface
    NUNC174922
    Log in for availability
    1 * 12 Items - 407,00 kr