Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH indicator, bromophenol blue tablets

pH indicator, bromophenol blue tablets

Bromophenol blue tablets, pH indicator, 100
Catalog # LOVI511620
Supplier:   Lovibond
pH indicator, bromophenol blue tablets
Catalog # LOVI511620
Supplier:  Lovibond
CAS Number:  
pH indicator, bromophenol blue tablets
pH indicator, bromophenol blue tablets

Bromophenol blue tablets, pH indicator, 100

CAS Number:  
Price
442,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Indicator for pH range 2,8 to 4,4.

Can be used for measuring pH in conjunction with the Comparator 2000+ system and pH disc 2/1B

Specifications

Specifications

  • Pakn:
    100
  • Beskrivelse:
    Bromophenol blue tablets, pH indicator

Specifications