Order Entry
Denmark
Orders LinkContactUsLinkComponent
Storage Bags
Storage Bags
255×355 mm
Catalog # 300-0046
Supplier:   Alpha Packaging
Storage Bags
Catalog # 300-0046
Supplier:  Alpha Packaging
Storage Bags
Storage Bags
255×355 mm
Price
274,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
Heavy duty storage bags, 500 gauge. Suitable for heat sealing. Clear PE, plain.
Specifications
  • Dimensions:
    255×355 mm
  • Seq:
    02