Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for LABmax, MINISPENSOR, TITREX

Accessories for LABmax, MINISPENSOR, TITREX

Discharge tube, bent, volume 2,5 - 10 ml, length 80 ml
Catalog # WITG5.375.003
Supplier:   witeg Labortechnik
Accessories for LABmax, MINISPENSOR, TITREX
Catalog # WITG5.375.003
Supplier:  witeg Labortechnik
Accessories for LABmax, MINISPENSOR, TITREX
Accessories for LABmax, MINISPENSOR, TITREX

Discharge tube, bent, volume 2,5 - 10 ml, length 80 ml

Price
96,10 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Discharge tube, bent, volume 2,5 - 10 ml, length 80 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.