Order Entry
Denmark
Orders LinkContactUsLinkComponent
WHEATON® Rubber Stoppers

WHEATON® Rubber Stoppers

Stopper, grey chlorobutyl, 7,0 mm, 13,0 mm
Catalog # 215-4716
Supplier:   DWK Life Sciences
WHEATON® Rubber Stoppers
Catalog # 215-4716
Supplier:  DWK Life Sciences
WHEATON® Rubber Stoppers
WHEATON® Rubber Stoppers

Stopper, grey chlorobutyl, 7,0 mm, 13,0 mm

Price
3.030,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Stoppers of various styles, sizes and rubber formulations.

  • Manufactured from the highest quality raw materials
  • Components manufactured to yield exceptional dimensional stability
  • Autoclavable

Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.

Snap-on style provides maximum sealing for vials with a blow-back glass finish.

Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.

Specifications
  • Ø bottom:
    7,0 mm
  • Ø top:
    13,0 mm
  • Description:
    Stopper, grey chlorobutyl
  • Seq:
    18
  • Cat. No.:
    215-4716
  • Durometer hardness:
    55