Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Manual valve gas and vacuum tubing connection kit, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Catalog # LBCP5245100
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5245100
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Manual valve gas and vacuum tubing connection kit, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Price
2.260,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Manual valve gas and vacuum tubing connection kit, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
  • Seq:
    02