Accessories for glove boxes, Precise™
Add-A-Valve kit, incl. manual ball valve, compression fitting, hose connector fitting, O-ring and nut to add a service valve to the main chamber or transfer chamber; requires installation and Ø 0,688" hole
Catalog # LBCP5225701
Supplier: LABCONCO
Gå til:
Product Details & Documents
Specifikationer
- Description:Add-A-Valve kit, incl. manual ball valve, compression fitting, hose connector fitting, O-ring and nut to add a service valve to the main chamber or transfer chamber; requires installation and Ø 0,688" hole
- Seq:05