Order Entry
Denmark
Orders LinkContactUsLinkComponent
Quality control test suspensions

Quality control test suspensions

Staphylococcus aureus, 2,5 mL
Katalog # NAMSGP25-08
Leverandør:   CROSSTEX
Quality control test suspensions
Katalog # NAMSGP25-08
Leverandør:  CROSSTEX
CAS nummer:  
Quality control test suspensions

Staphylococcus aureus, 2,5 mL

CAS nummer:  
Price
1.930,00 kr
Log ind for tilgængelighed
1 * 2,5 mL
Product Details & Documents

The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.

  • 2,5 ml pro vial
  • 25 dose
Specifikationer

Specifikationer

  • Pk:
    2,5 mL
  • Description:
    Staphylococcus aureus

Specifikationer