Order Entry
Denmark
Orders LinkContactUsLinkComponent
Quality control test suspensions

Quality control test suspensions

Aspergillus brasiliensis, 10 mL
Catalog # NAMSGP-04
Supplier:   CROSSTEX
Quality control test suspensions
Catalog # NAMSGP-04
Supplier:  CROSSTEX
CAS Number:  
Quality control test suspensions

Aspergillus brasiliensis, 10 mL

CAS Number:  
Price
4.740,00 kr
Log in for availability
1 * 10 mL
Product Details & Documents

The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.

  • 10 ml pro vial
  • 100 dose

Documents

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    Aspergillus brasiliensis

Specifications