Order Entry
Denmark
Orders LinkContactUsLinkComponent
ICP-MS interference check mixes

ICP-MS interference check mixes

6020 Interference check solution B. For use with ICP-MS systems. Contains: 20 ppm of Cr, Co, Cu, Mn, Ni and V, 10 ppm of As, Cd, Se, and Zn, 5 ppm of Ag. Bottle of 100 ml, 5% HNO₃, 1
Catalog # HEWL5188-6527
ICP-MS interference check mixes
Catalog # HEWL5188-6527
CAS Number:  
ICP-MS interference check mixes
ICP-MS interference check mixes

6020 Interference check solution B. For use with ICP-MS systems. Contains: 20 ppm of Cr, Co, Cu, Mn, Ni and V, 10 ppm of As, Cd, Se, and Zn, 5 ppm of Ag. Bottle of 100 ml, 5% HNO₃, 1

CAS Number:  
Price
2.320,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

It offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for atomic spectroscopy.

  • It is works in AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations

Standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    1
  • Description:
    6020 Interference check solution B. For use with ICP-MS systems. Contains: 20 ppm of Cr, Co, Cu, Mn, Ni and V, 10 ppm of As, Cd, Se, and Zn, 5 ppm of Ag. Bottle of 100 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.