VWR®, Vials til opbevaring
Clear glass, 20,00 ml, 22,7×86 mm, klar, No
Catalog # 548-2218
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Vials med skruelåg, i klart eller brunt glas i 1. hydrolytiske klasse til prøveopbevaring. Et bredt udvalg af skruelåg med lukket top fås med forskellige typer septum.
Specifications
- Kasket inkluderet:No
- Diameter×H:22,7×86 mm
- Model:Clear glass
- Kapacitet:20,00 ml
- Farve:klar
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



