Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Methylheptane ≥99%

2-Methylheptane ≥99%

Glass bottle for solids, 25 g
Catalog # H26220.14
Flammable
Health
Irritant
Environment
2-Methylheptane ≥99%
Catalog # H26220.14
CAS Number:  592-27-8
2-Methylheptane ≥99%
2-Methylheptane ≥99%

Glass bottle for solids, 25 g

CAS Number:  592-27-8
Price
9.820,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 592-27-8 Molecular Formula: C8H18 Formula Weight: 114.23 Flash Point: 4°(40°F) Refractive Index: 1.3950 Boiling Point: 116° Beilstein Reference: 1696862 UN No.: 1262 MDL No.: MFCD00009518

Synonyms:  Isoheptane
  • CAS Number:592-27-8
  • Formula:C₈H₁₈
  • MDL Number:MFCD00009518
  • Molecular Weight:114.23 g/mol
  • Boiling Pt:116 °C
  • Melting Pt:–109 °C
  • Density:0.698
  • Flash Pt:4 °C (40 °F)
  • Storage Temperature:Ambient
  • EINECS:209-747-9
  • UN:1262
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.