Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Methylheptane ≥99%

2-Methylheptane ≥99%

Glass bottle for solids, 5 g
Catalog # H26220.06
Flammable
Health
Irritant
Environment
2-Methylheptane ≥99%
Catalog # H26220.06
CAS Number:  592-27-8
2-Methylheptane ≥99%
2-Methylheptane ≥99%

Glass bottle for solids, 5 g

CAS Number:  592-27-8
Price
2.660,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 592-27-8 Molecular Formula: C8H18 Formula Weight: 114.23 Flash Point: 4°(40°F) Refractive Index: 1.3950 Boiling Point: 116° Beilstein Reference: 1696862 UN No.: 1262 MDL No.: MFCD00009518

Synonyms:  Isoheptane
  • CAS Number:592-27-8
  • Formula:C₈H₁₈
  • MDL Number:MFCD00009518
  • Molecular Weight:114.23 g/mol
  • Boiling Pt:116 °C
  • Melting Pt:–109 °C
  • Density:0.698
  • Flash Pt:4 °C (40 °F)
  • Storage Temperature:Ambient
  • EINECS:209-747-9
  • UN:1262
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications