Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stearic acid ≥90%

Stearic acid ≥90%

Glass bottle for solids, 5.000 g
Catalog # A17673.0I
Irritant
Stearic acid ≥90%
Catalog # A17673.0I
CAS Number:  57-11-4
Stearic acid ≥90%
Stearic acid ≥90%

Glass bottle for solids, 5.000 g

CAS Number:  57-11-4
Price
1.150,00 kr
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 90+% CAS No.: 57-11-4 Molecular Formula: CH3(CH2)16CO2H Formula Weight: 284.48 Flash Point: 196°(384°F) Odour: Odourless Boiling Point: 232°/15mm Beilstein Reference: 608585 Merck Reference: 14,8804 MDL No.: MFCD00002752

Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:14,8804
  • Boiling Pt:232 °C (15 mmHg)
  • Melting Pt:67…70 °C
  • Density:0.941
  • Flash Pt:196 °C (384 °F)
  • Storage Temperature:Ambient
  • EINECS:200-313-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.