Aluminon
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 569-58-4 Molecular Formula: C22H23N3O9 Formula Weight: 473.44 Odour: Odourless pH: 5.3 Melting Point: 220-225° dec. MDL No.: MFCD00040925
Synonyms: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
- CAS Number:569-58-4
- Formula:C₂₂H₂₃N₃O₉
- MDL Number:MFCD00040925
- Molecular Weight:473.44 g/mol
- Merck Index:14,322
- Melting Pt:220…225 °C (dec.)
- Storage Temperature:Ambient
- EINECS:209-319-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



