opus dispenser, with reagent recirculation-system, conformity certified, serial or USB interface, for dispenser and titration 1 * 1 items
Catalog # 613-0758
Leverandør: HIRSCHMANN
opus dispenser, with reagent recirculation-system, conformity certified, serial or USB interface, for dispenser and titration 1 * 1 items
Catalog # 613-0758
Leverandør: HIRSCHMANN
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
VWR®, Nitrile Gloves
Leverandør: VWR Collection
VWR® Facial Tissues
Leverandør: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Leverandør: Merck
Sealing film for manual application, Parafilm® M
Leverandør: HEATHROW
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Leverandør: VWR Collection
VWR®, Transfer Pipettes, High Performance
Leverandør: VWR Collection
VWR® Serological Pipette, Standard Line
Leverandør: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Leverandør: KIMBERLY CLARK