Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Acenaphthene-[D₁₀] (99% D)

Glass bottle for solids, 1 g
Catalog # CAMBDLM-108-1
Environment
Acenaphthene-[D₁₀] (99% D)
Catalog # CAMBDLM-108-1
CAS Number:  15067-26-2
Acenaphthene-[D₁₀] (99% D)

Glass bottle for solids, 1 g

CAS Number:  15067-26-2
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Acenaphthene-[²H₁₀]
  • CAS Number:15067-26-2
  • Formula:C₁₂D₁₀
  • Molecular Weight:164.13 g/mol
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.