Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Blue
Catalog # 720-1439
Supplier: Simport Scientific
Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Catalog # 720-1439
Supplier: Simport Scientific
Jump to:
Product Details & Documents
To be used with Leica and Sakura inkjet printers.
- The one-piece disposable plastic cover is pre-installed on each cassette
- Eliminates the need for reusable steel lids
- The cover can be opened and closed as often as necessary and will always relock
Moulded from a special high density acetal polymer, they keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage. The cover does not protrude above the cassette, a great space saving feature.
Packaging: Cassettes are packed in stacks of 40. Each case contains 50 stacks for a total of 2000 cassettes.
Specifications
- Colour:Blue
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



