Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Green
Katalog # 720-1435
Leverandør: Simport Scientific
Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Katalog # 720-1435
Leverandør: Simport Scientific
Gå til:
Product Details & Documents
To be used with Leica and Sakura inkjet printers.
- The one-piece disposable plastic cover is pre-installed on each cassette
- Eliminates the need for reusable steel lids
- The cover can be opened and closed as often as necessary and will always relock
Moulded from a special high density acetal polymer, they keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage. The cover does not protrude above the cassette, a great space saving feature.
Packaging: Cassettes are packed in stacks of 40. Each case contains 50 stacks for a total of 2000 cassettes.
Specifikationer
- Colour:Green



