Order Entry
Denmark
ContactUsLinkComponent
Ph-combination electrode with 1 m fixed cable and bnc plug, glass shaft, ceramic diaphragm, electrolyte kcl 3 mol/l, iodine/iodide-r eference system, sphere membrane, a-glass, length 170 mm, 12 mm , -5...+100 c, 0...14 ph 1 * 1 items
Catalog # 662-6079
Supplier:  SI Analytics
Ph-combination electrode with 1 m fixed cable and bnc plug, glass shaft, ceramic diaphragm, electrolyte kcl 3 mol/l, iodine/iodide-r eference system, sphere membrane, a-glass, length 170 mm, 12 mm , -5...+100 c, 0...14 ph 1 * 1 items
Catalog # 662-6079
Supplier:  SI Analytics
Frequently Bought Together