Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tripods

Tripods

Tripod, outer Ø 140 mm, 110×280 mm, LABOGAZ® Bunsen burners No. 41395010 and 41396010
Catalog # HECH41091010
Tripods
Catalog # HECH41091010
Tripods
Tripods

Tripod, outer Ø 140 mm, 110×280 mm, LABOGAZ® Bunsen burners No. 41395010 and 41396010

Price
224,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These tripods are made of galvanised iron.

Specifications
  • For:
    LABOGAZ® Bunsen burners No. 41395010 and 41396010
  • Int.Ø×H:
    110×280 mm
  • Description:
    Tripod, outer Ø 140 mm
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    Pk Info
    Pk
    Availability
    Price
  • HECH51071012
    120x120 mm
    1
    Log in for availability
    1 * 1 Items - 28,70 kr