Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tripods

Tripods

Tripod, outer Ø 140 mm, 110×210 mm, -
Catalog # HECH41090010
Tripods
Catalog # HECH41090010
Tripods
Tripods

Tripod, outer Ø 140 mm, 110×210 mm, -

Price
184,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These tripods are made of galvanised iron.

Specifications
  • For:
    -
  • Int.Ø×H:
    110×210 mm
  • Description:
    Tripod, outer Ø 140 mm
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Pk Info
    Pk
    Availability
    Price
  • HECH51071012
    120x120 mm
    1
    Log in for availability
    1 * 1 Items - 28,70 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.