Order Entry
Denmark
Orders LinkContactUsLinkComponent
Transfer pipettes, fine tip

Transfer pipettes, fine tip

Small bulb fine tip, 0,5 ml, 25 µl, 36 mm, -, Bulk
Catalog # ALPCLW4697
Supplier:   Alpha Laboratories
Transfer pipettes, fine tip
Catalog # ALPCLW4697
Supplier:  Alpha Laboratories
Transfer pipettes, fine tip
Transfer pipettes, fine tip

Small bulb fine tip, 0,5 ml, 25 µl, 36 mm, -, Bulk

Price
1.030,00 kr
Log in for availability
1 * 500 Items
Product Details & Documents

Non toxic, food grade LDPE.

  • For careful control and small volume transfers
  • Ideal for supernatant removal, slide preparation and gel loading
Specifications
  • Packed:
    Bulk
  • Sterile:
    -
  • Capacity:
    0,5 ml
  • Description:
    Small bulb fine tip
  • Length:
    36 mm
  • Seq:
    01
  • Drop vol.:
    25 µl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.